Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH3BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530483
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (88%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.
Specifications
SH3BP1 | |
Polyclonal | |
Unconjugated | |
SH3BP1 | |
3bp1; 3BP-1; ARHGAP43; OTTHUMP00000028981; SH3 domain binding protein 1; SH3 domain-binding protein 1; SH3BP1; SH3-domain binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20401, 23616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P55194, Q9Y3L3 | |
SH3BP1 | |
Recombinant protein encompassing a sequence within the center region of human SH3BP1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction