Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581891 Supplier Invitrogen™ Supplier No. PA581891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENLSNLRYLM KFLARLAEEQ EVNKMTPSNI AIVLGPNLLW PPEKEGDQAQ LDAASVSSIQ VVGVVEALIQ SADTLFPGDI NFNVSGLFSA VTLQDTVSDR LASEELPSTA VPTPATTPAP APAPAPAPAP ALASAATKER TESEVPP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 75%.

Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3BP1
Gene Accession No. Q9Y3L3
Gene Alias 3bp1; 3BP-1; ARHGAP43; OTTHUMP00000028981; SH3 domain binding protein 1; SH3 domain-binding protein 1; SH3BP1; SH3-domain binding protein 1
Gene Symbols SH3BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3BP1. Recombinant protein control fragment (Product #RP-91341).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23616
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.