Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3BP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158872
Description
SH3BP2 Polyclonal specifically detects SH3BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SH3BP2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Abl-SH3 binding protein 2, Cherubism, CRBM, CRPM, FLJ42079, FLJ54978,3BP-2,3BP2, RES4-23, SH3 domain-binding protein 2, SH3-domain binding protein 2, TNFAIP3 interacting protein 2 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Rat: 100%; Guinea pig: 92%; Rabbit: 92%; Bovine: 84%; Chicken: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P78314 | |
SH3BP2 | |
Synthetic peptides corresponding to SH3BP2(SH3-domain binding protein 2) The peptide sequence was selected from the middle region of SH3BP2. Peptide sequence RQPSQADTGGDDSDEDYEKVPLPNSVFVNTTESCEVERLFKATSPRGEPQ. | |
100 μL | |
Cell Biology, Signal Transduction | |
6452 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction