Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3BP2, p Ser427 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600492
Description
SH3BP2 Polyclonal specifically detects SH3BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
SH3BP2 (p Ser427) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
Abl-SH3 binding protein 2, Cherubism, CRBM, CRPM, FLJ42079, FLJ54978,3BP-2,3BP2, RES4-23, SH3 domain-binding protein 2, SH3-domain binding protein 2, TNFAIP3 interacting protein 2 | |
Rabbit | |
62.24 kDa | |
0.1 mg | |
Cell Biology, Phospho Specific, Signal Transduction | |
6452 | |
Reconstitute with 50 ul of distilled water to a final concentration of 1.0 mg/ml. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:2000-1:10000, Immunoprecipitation 1:100 | |
P78314 | |
SH3BP2 | |
This affinity purified SH3BP2 [p Ser427] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 415-440 of Human SH3BP2 protein (SH3 Domain Binding Protein 2). (Uniprot: P78314) | |
Affinity Purified | |
RUO | |
Primary | |
This product is an affinity purified antibody produced by immunoaffinity chromatography and is phospho specific for human SH3BP2. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction