Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH3BP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587739
Description
Immunogen sequence: VPEAMARPAV LPRPEKPQLP HLQRSPPDGQ SFRSFSFEKP RQPSQADTGG DDSDEDYEKV PLPNSVFVNT TESCEVERLF KATSPRGEPQ DGLYCIRNSS TKSGKVLVVW DETSNKVRNY RIFEKDSKFY LEGEVLFVSV GSMVEHYHTH VLPSHQSLLL RHPYGYTGPR; Positive Samples: A549, Mouse spleen, Mouse lung.
The protein encoded by this gene has an N-terminal pleckstrin homology (PH) domain, an SH3-binding proline-rich region, and a C-terminal SH2 domain. The protein binds to the SH3 domains of several proteins including the ABL1 and SYK protein tyrosine kinases, and functions as a cytoplasmic adaptor protein to positively regulate transcriptional activity in T, natural killer (NK), and basophilic cells. Mutations in this gene result in cherubism. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
SH3BP2 | |
Polyclonal | |
Unconjugated | |
Sh3bp2 | |
3BP2; 3BP-2; Abl-SH3 binding protein 2; CRBM; CRPM; FLJ42079; LOW QUALITY PROTEIN: SH3 domain-binding protein 2; RES4-23; SH3 domain binding protein 2; SH3 domain-binding protein 2; SH3BP2; SH3-domain binding protein 2; TNFAIP3 interacting protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24055, 6452 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78314, Q06649 | |
Sh3bp2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 392-561 of human SH3BP2 (NP_0011161531). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction