Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3BP4 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177031MFV450
Description
SH3BP4 Polyclonal antibody specifically detects SH3BP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SH3BP4 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BOG25SH3 domain-binding protein 4, EHB10, EH-binding protein 10, SH3-domain binding protein 4, transferrin receptor trafficking protein, Transferrin receptor-trafficking protein, TTP | |
Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human SH3BP4. The immunogen is located within amino acids 860 - 910 of SH3BP4. Amino Acid Squence: MDAYESPHRDRNGVVDSE | |
0.1 mL | |
Cell Cycle and Replication | |
23677 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction