Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557992 Supplier Invitrogen™ Supplier No. PA557992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGTLIDLSEG FSETSFNDIK VPSPSALLVD NPTPFGNAKE VIAIKDYCPT NFTTLKFSKG DHLYVLDTSG GEWWYAHNT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

SH3BP4 was initially isolated as a novel cDNA using differential display analysis using cultured human corneal fibroblasts. The protein contains three Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. SH3BP4 has been shown to interact with specific endocytic proteins such as clathrin, dynamin, and the transferrin receptor (TfR) and localizes to TfR-containing coated pits and vesicles. It is thought to be involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of TfR. At least two isoforms of SH3BP4 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3BP4
Gene Accession No. Q9P0V3
Gene Alias AI594717; AW227605; BOG25; EHB10; EH-binding protein 10; SH3 domain binding protein 4; SH3 domain-binding protein 4; SH3 domain-binding protein 4; SH3-domain binding protein 4; Sh3bp4; SH3-domain binding protein 4; si:dkey-57b6.3; Transferrin receptor-trafficking protein; TTP
Gene Symbols SH3BP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3BP4. Recombinant protein control fragment (Product #RP-96078).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.