Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BP4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572206
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572206 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572206 Supplier Invitrogen™ Supplier No. PA572206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH3BP4A Polyclonal Antibody for Western Blot

Possible role in regulating endocytosis of the transferrin receptor at the plasma membrane. Alternatively, may function as a negative regulator of the amino acid-induced TOR signaling by inhibiting the formation of active Rag GTPase complexes. Preferentially binds inactive Rag GTPase complexes and prevents their interaction with the mTORC1 complex inhibiting its relocalization to lysosomes and its activation. Thereby, may indirectly regulate cell growth, proliferation and autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BP4A
Applications Western Blot
Classification Polyclonal
Concentration 2.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene sh3bp4a
Gene Accession No. Q1LVQ2
Gene Alias SH3 binding protein-4; SH3 domain-binding protein 4-A; sh3bp4; sh3bp4 protein; sh3bp4a; SH3-domain binding protein 4; SH3-domain binding protein 4a; si:dkey-110p14.1; src homology 3 binding protein 4; truncated SH3 binding domain protein 4
Gene Symbols sh3bp4a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 132-165 amino acids from the N-terminal region of DANRE sh3bp4a.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403082
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.