Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3BP5 Polyclonal Antibody, Abbomax

Catalog No. 502204590
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4590 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4590 Supplier Abbomax Inc Supplier No. 630490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Ovarian cancer Cellular location: Mitochondria.

SH3BP5 Polyclonal antibody specifically detects SH3BP5 in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SH3BP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene SH3BP5
Gene Accession No. O60239, Q91Y80, Q9Z131
Gene Alias AI606498; Sab; SH3 binding protein; SH3 domain binding protein 5; SH3 domain-binding protein 5; SH3 domain-binding protein that preferentially associates with BTK; SH3BP5; SH3BP-5; SH3-domain binding protein 5 (BTK-associated); Vascular endothelial cell-specific protein 18; Vesp18
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of rat DCLK1
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117186, 24056, 618056, 9467
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.