Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3BP5L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | SH3BP5L |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SH3BP5L Polyclonal specifically detects SH3BP5L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SH3BP5L | |
Polyclonal | |
Rabbit | |
Human | |
Q7L8J4 | |
80851 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TQGSHLGSCIEKARPYYEARRLAKEAQQETQKAALRYERAVSMHNAAREMVFVAEQGVMADKNRLDPTWQEMLNHATCK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ33845, KIAA1720SH3BP-5-like, SH3 domain-binding protein 5-like, SH3-binding domain protein 5-like | |
SH3BP5L | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title