Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3D19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566183 Supplier Invitrogen™ Supplier No. PA566183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAEESVGSEMV LDPFQLPAKT EPIKERAVQP APTRKPTVIR IPAKPGKCLH EDPQSPPPLP AEKPIGNTFS TVSGKLSN Highest antigen sequence identity to the following orthologs - mouse 82%, rat 84%.

May play a role in regulating A disintegrin and metalloproteases (ADAMs) in the signaling of EGFR-ligand shedding. May be involved in suppression of Ras-induced cellular transformation and Ras-mediated activation of ELK1. Plays a role in the regulation of cell morphology and cytoskeletal organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3D19
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3D19
Gene Accession No. Q5HYK7
Gene Alias ADAM-binding protein Eve-1; AW011754; EBP; EEN-binding protein; EVE1; Kryn; RGD1304885; SH3 domain containing 19; SH3 domain containing protein D 19; SH3 domain protein D19; SH3 domain-containing protein 19; SH3D19; SH3P19
Gene Symbols SH3D19
Host Species Rabbit
Immunogen Recombinant Human SH3D19. Recombinant protein control fragment (Product #RP-106870).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.