Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3D19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570805 Supplier Thermo Scientific Supplier No. PA570805
Add to Cart
Add to Cart

SH3D19 Polyclonal Antibody

SH3D19 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3D19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SH3D19
Gene Accession No. Q5HYK7
Gene Alias ADAM-binding protein Eve-1; AW011754; EBP; EEN-binding protein; EVE1; Kryn; RGD1304885; SH3 domain containing 19; SH3 domain containing protein D 19; SH3 domain protein D19; SH3 domain-containing protein 19; SH3D19; SH3P19
Gene Symbols SH3D19
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human SH3D19.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152503
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.