Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH3GL1 Monoclonal Antibody (2A9H4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538630
Description
This antibody has been tested in indirect ELISA.
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
SH3GL1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q99961 | |
SH3GL1 | |
Purified recombinant fragment of human SH3GL1 (aa: 12-119) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
2A9H4 | |
Unconjugated | |
SH3GL1 | |
C77078; CNSA1; EEN; EEN fusion partner of MLL; endophilin A2; endophilin II; endophilin-2; Endophilin-A2; extra 11-19 leukemia fusion; extra eleven-nineteen leukemia fusion gene protein; MGC111371; SH3 domain containing GRB2 like 1; SH3 domain containing GRB2 like 1, endophilin A2; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3-containing Grb-2-like 1 protein; Sh3d2b; SH3-domain GRB2-like 1; SH3GL1; Sh3p8 | |
Mouse | |
Protein G | |
RUO | |
6455 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction