Learn More
Invitrogen™ SH3GL1 Monoclonal Antibody (OTI2F5)

Description
SH3GL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | SH3GL1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2F5 |
| Concentration | 0.64 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SH3GL1 |
| Gene Accession No. | Q99961 |
| Gene Alias | C77078; CNSA1; EEN; EEN fusion partner of MLL; endophilin A2; endophilin II; endophilin-2; Endophilin-A2; extra 11-19 leukemia fusion; extra eleven-nineteen leukemia fusion gene protein; MGC111371; SH3 domain containing GRB2 like 1; SH3 domain containing GRB2 like 1, endophilin A2; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3-containing Grb-2-like 1 protein; Sh3d2b; SH3-domain GRB2-like 1; SH3GL1; Sh3p8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.