Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3GL1 Monoclonal Antibody (OTI3F8)
GREENER_CHOICE

Catalog No. PIMA525202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525202 Supplier Invitrogen™ Supplier No. MA525202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3GL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3GL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3F8
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH3GL1
Gene Accession No. Q99961
Gene Alias C77078; CNSA1; EEN; EEN fusion partner of MLL; endophilin A2; endophilin II; endophilin-2; Endophilin-A2; extra 11-19 leukemia fusion; extra eleven-nineteen leukemia fusion gene protein; MGC111371; SH3 domain containing GRB2 like 1; SH3 domain containing GRB2 like 1, endophilin A2; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3-containing Grb-2-like 1 protein; Sh3d2b; SH3-domain GRB2-like 1; SH3GL1; Sh3p8
Gene Symbols SH3GL1
Host Species Mouse
Immunogen Full length human recombinant protein of SH3GL1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6455
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.