Learn More
Invitrogen™ SH3GL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120939
Description
Positive test controls include: HeLa, BT-474, 293T, HepG2, Mouse brain, Mouse testis. The target is usually found in the following locations: Cytoplasm, Membrane, Peripheral membrane protein. Immunogen sequence: MSVAGLKKQF HKATQKVSEK VGGAEGTKLD DDFKEMERKV DVTSRAVMEI MTKTIEYLQP NPASRAKLSM INTMSKIRGQ EKGPGYPQAE ALLAEAMLKF GRELGDDCNF GPALGEVGEA MRELSEVKDS LDIEVKQNFI DPLQNLHDKD LREIQHHLKK LEGRRLDFDY KKKRQGKIPD EELRQALEKF DESKEIAESS MFNLLEMDIE QVSQLSALVQ AQLEYHKQAV QILQQVTVRL EERIRQASSQ PRREYQPKPR MSLEFPTGDS.
Enables identical protein binding activity. Involved in negative regulation of blood-brain barrier permeability; negative regulation of gene expression; and negative regulation of protein phosphorylation. Located in perinuclear region of cytoplasm.
Specifications
| SH3GL2 | |
| Polyclonal | |
| Unconjugated | |
| Sh3gl2 | |
| 9530001L19Rik; AI120490; AW555077; B930049H17Rik; bA335L15.1; bA335L15.1 (SH3-domain GRB2-like 2); CNSA2; Een1; EEN-B1; endophilin; endophilin A1; Endophilin A1 BAR domain; endophilin I; endophilin-1; Endophilin-A1; FLJ20276; FLJ25015; SH3 domain containing GRB2 like 2; SH3 domain containing GRB2 like 2, endophilin A1; SH3 domain protein 2A; SH3 domain-containing GRB2-like protein 2; Sh3d2a; SH3-domain GRB2-like 2; SH3GL2; Sh3p4; SH3PA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20404, 6456 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62420, Q99962 | |
| Sh3gl2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SH3GL2 (NP_003017.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.