Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3GL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555377 Supplier Invitrogen™ Supplier No. PA555377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALVQAQLEYH KQAVQILQQV TVRLEERIRQ ASSQPRREYQ PKPRMSLEFP TGDSTQPNGG LSHTGTPKPS GVQMDQPCCR ALYDFEPENE GELG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

Enables identical protein binding activity. Involved in negative regulation of blood-brain barrier permeability; negative regulation of gene expression; and negative regulation of protein phosphorylation. Located in perinuclear region of cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3GL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh3gl2
Gene Accession No. Q99962
Gene Alias 9530001L19Rik; AI120490; AW555077; B930049H17Rik; bA335L15.1; bA335L15.1 (SH3-domain GRB2-like 2); CNSA2; Een1; EEN-B1; endophilin; endophilin A1; Endophilin A1 BAR domain; endophilin I; endophilin-1; Endophilin-A1; FLJ20276; FLJ25015; SH3 domain containing GRB2 like 2; SH3 domain containing GRB2 like 2, endophilin A1; SH3 domain protein 2A; SH3 domain-containing GRB2-like protein 2; Sh3d2a; SH3-domain GRB2-like 2; SH3GL2; Sh3p4; SH3PA
Gene Symbols Sh3gl2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3GL2. Recombinant protein control fragment (Product #RP-92910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.