Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3GL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118917 Supplier Invitrogen™ Supplier No. PA5118917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SH3GL3.

SH3GL3 is implicated in endocytosis. It may recruit other proteins to membranes with high curvature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3GL3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Sh3gl3
Gene Accession No. O35180, Q62421, Q99963
Gene Alias CNSA3; EEN-B2; Endophilin 3; endophilin A3; endophilin III; endophilin-3; Endophilin-A3; HsT19371; SH3 domain containing GRB2 like 3; SH3 domain containing GRB2 like 3, endophilin A3; SH3 domain containing GRB2 like endophilin A3; SH3 domain protein 2 C1; SH3 domain protein 2 C2; SH3 domain protein 2C; SH3 domain-containing GRB2-like protein 3; Sh3d2c; Sh3d2c1; Sh3d2c2; SH3-domain GRB2-like 3; Sh3gl3; SH3p13
Gene Symbols Sh3gl3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SH3GL3(Accession Q99963), corresponding to amino acid residues I77-V127.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20408, 6457, 81921
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.