Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3GLB2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297456AF350
Description
SH3GLB2 Polyclonal specifically detects SH3GLB2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SH3GLB2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
endophilin-B2, GRB2-like, endophilin B2, SH3-containing protein SH3GLB2, SH3-domain GRB2-like endophilin B2 | |
Produced in rabbits immunized with E. coli-derived Human SH3GLB2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
56904 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction