Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3GLB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555167 Supplier Invitrogen™ Supplier No. PA555167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDFVKSQTTY YAQCYRHMLD LQKQLGRFPG TFVGTTEPAS PPLSSTSPTT AAATMPVVPS VASLAPPGEA SLCLEEVAPP ASGTRKARVL Y Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 86%.

SH3GLB2 (SH3 Domain Containing GRB2 Like, Endophilin B2) is a Protein Coding gene. Among its related pathways are Endocytosis. An important paralog of this gene is SH3GLB1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3GLB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3GLB2
Gene Accession No. Q9NR46
Gene Alias EMBL:AAH87652.1}; endophilin-B2; Kiaa1848; PP578; PP6569; PP9455; retinoid receptor-induced gene 1; RRIG1; SH3 domain containing GRB2 like endophilin B2; SH3 domain-containing GRB2-like protein B2; SH3-containing protein SH3GLB2; SH3-domain GRB2-like endophilin B2; Sh3glb2; sh3glb2 {ECO:0000312; SPAS-1; stimulator of prostatic adenocarcinoma-specific T-cells-1
Gene Symbols SH3GLB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3GLB2. Recombinant protein control fragment (Product #RP-93013).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.