Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3KBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118261 Supplier Invitrogen™ Supplier No. PA5118261
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SH3KBP1 Polyclonal Antibody for Western Blot, IHC (P), IP

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3KBP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Sh3kbp1
Gene Accession No. Q96B97
Gene Alias 1200007H22Rik; 1700125L08Rik; 5830464D22Rik; AI447724; cbl-interacting protein of 85 kDa; c-Cbl-interacting protein; CD2-binding protein 3; CD2BP3; CIN85; GIG10; HSB1; HSB-1; Human Src family kinase-binding protein 1; IN85; LOW QUALITY PROTEIN: SH3 domain-containing kinase-binding protein 1; MIG18; migration-inducing gene 18; regulator of ubiquitous kinase; Ruk; Seta; SH3 domain containing kinase binding protein 1; SH3 domain-containing adapter protein; SH3 domain-containing kinase-binding protein 1; SH3-containing, expressed in tumorigenic astrocytes; SH3-domain kinase binding protein 1; Sh3kbp1; Src family kinase-binding protein 1; src-related kinase binding protein-1
Gene Symbols Sh3kbp1
Host Species Rabbit
Immunogen E. coli-derived Human SH3KBP1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.