Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3KBP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984015
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4015 Supplier Bioss Supplier No. BS16723R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.

Specifications

Antigen SH3KBP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Sh3kbp1
Gene Accession No. Q96B97
Gene Alias 1200007H22Rik; 1700125L08Rik; 5830464D22Rik; AI447724; BOS_26313; Cbl-interacting protein of 85 kDa; c-Cbl-interacting protein; CD2-binding protein 3; CD2BP3; CIN85; GIG10; HSB1; HSB-1; Human Src family kinase-binding protein 1; IN85; LOW QUALITY PROTEIN: SH3 domain-containing kinase-binding protein 1; MIG18; migration-inducing gene 18; Regulator of ubiquitous kinase; Ruk; Seta; SH3 domain containing kinase binding protein 1; SH3 domain-containing adapter protein; SH3 domain-containing kinase-binding protein 1; SH3-containing, expressed in tumorigenic astrocytes; SH3-domain kinase binding protein 1; Sh3kbp1; Src family kinase-binding protein 1; src-related kinase binding protein-1
Gene Symbols Sh3kbp1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SH3KBP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.