Learn More
SH3PXD2A Mouse anti-Human, Clone: OTI1F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811757

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.Specifications
SH3PXD2A | |
Monoclonal | |
Unconjugated | |
SH3PXD2A | |
FISH, SH3MD1, TKS5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5TCZ1 | |
SH3PXD2A | |
Human recombit protein fragment corresponding to amino acids 299-395 of human SH3PXD2A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.