Learn More
Invitrogen™ SH3PXD2A Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Specifications
Specifications
| Antigen | SH3PXD2A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Sh3pxd2a |
| Gene Accession No. | Q5TCZ1 |
| Gene Alias | 2310014D11Rik; AA589508; Adapter protein TKS5; adaptor protein TKS5; AI256723; AI413738; C230050L11; EG329070; FISH; Five SH3 domain-containing protein; five SH3 domains; Gm5098; KIAA0418; SH3 and PX domain-containing protein 2A; SH3 and PX domains 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Sh3md1; SH3PXD2A; Tks5; TSK5; Tyrosine kinase substrate with five SH3 domains |
| Gene Symbols | Sh3pxd2a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.