Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH3PXD2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116404
Description
SH3PXD2B Polyclonal Antibody for Western Blot
Adapter protein involved in invadopodia and podosome formation and extracellular matrix degradation. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. Plays a role in mitotic clonal expansion during the immediate early stage of adipocyte differentiation.
Specifications
SH3PXD2B | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
A1X283 | |
SH3PXD2B | |
KLH conjugated synthetic peptide between 505-539 amino acids from the Central region of human SH3PXD2B. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SH3PXD2B | |
adapter protein HOFI; adaptor protein HOFI; factor for adipocyte differentiation 49; FAD49; FLJ20831; FTHS; HOFI; KIAA1295; SH3 and PX domain-containing protein 2B; SH3 and PX domains 2B; SH3PXD2B; TKS4; TSK4; tyrosine kinase substrate with four SH3 domains | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
285590 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction