Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3PXD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557673 Supplier Invitrogen™ Supplier No. PA557673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPGVILPMMP AKHIPPARDS RRPEPKPDKS RLFQLKNDMG LECGHKVLAK EVKKPNLRPI SKSKTDLPEE KPDATPQNPF LKSRPQV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

Adapter protein involved in invadopodia and podosome formation and extracellular matrix degradation. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. Plays a role in mitotic clonal expansion during the immediate early stage of adipocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3PXD2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3PXD2B
Gene Accession No. A1X283
Gene Alias adapter protein HOFI; adaptor protein HOFI; factor for adipocyte differentiation 49; FAD49; FLJ20831; FTHS; HOFI; KIAA1295; SH3 and PX domain-containing protein 2B; SH3 and PX domains 2B; SH3PXD2B; TKS4; TSK4; tyrosine kinase substrate with four SH3 domains
Gene Symbols SH3PXD2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3PXD2B. Recombinant protein control fragment (Product #RP-96944).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.