Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3RF1 Monoclonal Antibody (1F7)
GREENER_CHOICE

Catalog No. PIMA522392
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522392 Supplier Invitrogen™ Supplier No. MA522392
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LAQDAFHRKA SSLDSAVPIA PPPRQACSSL GPVLNESRPV VCERHRVVVS YPPQSEAELE LKEGDIVFVH KKREDGWFKG TLQRNGKTGL FPGSFVENI.

This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via the RING-finger, the encoded protein has been shown to function as an ubiquitin-protein ligase involved in protein sorting at the trans-Golgi network. The encoded protein may also act as a scaffold for the c-Jun N-terminal kinase signaling pathway, facilitating the formation of a functional signaling module.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SH3RF1
Gene Accession No. Q7Z6J0
Gene Alias 2200003J05Rik; E3 ubiquitin-protein ligase SH3RF1; FLJ21602; KIAA1494; plenty of SH3 domains; plenty of SH3s; Posh; Posh1; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative E3 ubiquitin-protein ligase SH3RF1; E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; R75531; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; RNF142; SH3 domain containing ring finger 1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2; SH3MD2; Sh3rf1
Gene Symbols SH3RF1
Host Species Mouse
Immunogen SH3RF1 (NP_065921, 790 a.a. ~ 888 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57630
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.