Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3RF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530779 Supplier Invitrogen™ Supplier No. PA530779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (90%), Rat (88%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via the RING-finger, the encoded protein has been shown to function as an ubiquitin-protein ligase involved in protein sorting at the trans-Golgi network. The encoded protein may also act as a scaffold for the c-Jun N-terminal kinase signaling pathway, facilitating the formation of a functional signaling module.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SH3RF1
Gene Accession No. Q7Z6J0
Gene Alias 2200003J05Rik; E3 ubiquitin-protein ligase SH3RF1; FLJ21602; KIAA1494; plenty of SH3 domains; plenty of SH3s; Posh; Posh1; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative E3 ubiquitin-protein ligase SH3RF1; E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; R75531; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; RNF142; SH3 domain containing ring finger 1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2; SH3MD2; Sh3rf1
Gene Symbols SH3RF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SH3MD2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.