Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3RF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563527 Supplier Invitrogen™ Supplier No. PA563527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGSLRKGRSS MRKNGSLQRP LQSGIPTLVV GSLRRSPTMV LRPQQFQFYQ PQGIPSSPSA VVVEMGSKPA LTGEPALTCI SRGSE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

The SH3RF2 gene encodes a protein that has been shown to possess protein phosphatase 1 binding activity and ubiquitin protein ligase activity. This protein is involved in several cellular processes, including the positive regulation of the JNK cascade, protein autoubiquitination, and regulation of cellular protein metabolic processes. The protein is located in the nucleoplasm and mutations in the SH3RF2 gene have been associated with Ureter Transitional Cell Carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3RF2
Gene Accession No. Q8TEC5
Gene Alias 2310046K19; 9130023G24Rik; AI452083; E3 ubiquitin-protein ligase SH3RF2; Heart protein phosphatase 1-binding protein; HEPP1; phosphatase 1 binding protein; Posh2; Posh3; POSH-eliminating RING protein; Posher; Ppp1r39; Protein phosphatase 1 regulatory subunit 39; protein phosphatase 1, regulatory subunit 39; putative E3 ubiquitin-protein ligase SH3RF2; RING finger protein 158; RING-type E3 ubiquitin transferase SH3RF2; Rnf158; SH3 domain containing ring finger 2; SH3 domain-containing RING finger protein 2; Sh3rf2
Gene Symbols SH3RF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3RF2. Recombinant protein control fragment (Product #RP-103317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.