Learn More
Invitrogen™ SH3RF2 Polyclonal Antibody

Description
A suggested positive control is NIH-3T3 cell lysate. PA5-21013 can be used with blocking peptide PEP-1127.
Specifications
Specifications
| Antigen | SH3RF2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SH3RF2 |
| Gene Accession No. | Q498M5, Q8BZT2, Q8TEC5 |
| Gene Alias | 2310046K19; 9130023G24Rik; AI452083; E3 ubiquitin-protein ligase SH3RF2; Heart protein phosphatase 1-binding protein; HEPP1; phosphatase 1 binding protein; Posh2; Posh3; POSH-eliminating RING protein; Posher; Ppp1r39; Protein phosphatase 1 regulatory subunit 39; protein phosphatase 1, regulatory subunit 39; putative E3 ubiquitin-protein ligase SH3RF2; RING finger protein 158; RING-type E3 ubiquitin transferase SH3RF2; Rnf158; SH3 domain containing ring finger 2; SH3 domain-containing RING finger protein 2; Sh3rf2 |
| Gene Symbols | SH3RF2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.