Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3RF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560841 Supplier Invitrogen™ Supplier No. PA560841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRNNVVGGSP LAKGITTTMH PGSGSLSSLA TATRPALPIT TPQAHAQHPT ASPPTGSCLR HSAQPTASQA RSTISTAAHS AAQAQDRPTA TVSPLRTQNS PSR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.

Has E3 ubiquitin-protein ligase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3RF3
Gene Accession No. Q8TEJ3
Gene Alias 4831416G18Rik; E3 ubiquitin-protein ligase SH3RF3; Igfbp5-ip; insulin-like growth factor binding protein 5 interacting protein; plenty of SH3s 2; plenty of SH3s-2; POSH2; SH3 domain containing ring finger 3; SH3 domain-containing RING finger protein 3; SH3 multiple domains 4; SH3 multiple domains protein 4; SH3MD4; Sh3rf3; Sh3rf31
Gene Symbols SH3RF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3RF3. Recombinant protein control fragment (Product #RP-99673).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.