Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3RF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101887 Supplier Invitrogen™ Supplier No. PA5101887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SH3RF3.

Has E3 ubiquitin-protein ligase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SH3RF3
Gene Accession No. Q8C120, Q8TEJ3
Gene Alias 4831416G18Rik; E3 ubiquitin-protein ligase SH3RF3; Igfbp5-ip; insulin-like growth factor binding protein 5 interacting protein; plenty of SH3s 2; plenty of SH3s-2; POSH2; SH3 domain containing ring finger 3; SH3 domain-containing RING finger protein 3; SH3 multiple domains 4; SH3 multiple domains protein 4; SH3MD4; Sh3rf3; Sh3rf31
Gene Symbols SH3RF3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SH3RF3(Accession Q8TEJ3), corresponding to amino acid residues K713-E763.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237353, 344558
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.