Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3TC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558258 Supplier Invitrogen™ Supplier No. PA558258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTVVMRPSVS WEKAGPEEAK APVRGDEAPP ARVAGPAAGT PPCQMGVYPT DLTLQLLAVR RKSRLRDPGL QQTLR Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

SH3TC1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3TC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3TC1
Gene Accession No. Q8TE82
Gene Alias SH3 domain and tetratricopeptide repeat-containing protein 1; SH3 domain and tetratricopeptide repeats 1; SH3 domain and tetratricopeptide repeats-containing protein 1; SH3TC1
Gene Symbols SH3TC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3TC1. Recombinant protein control fragment (Product #RP-94983).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.