Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3TC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104137 Supplier Invitrogen™ Supplier No. PA5104137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SH3TC2.

This gene encodes a protein with two N-terminal Src homology 3 (SH3) domains and 10 tetratricopeptide repeat (TPR) motifs, and is a member of a small gene family. The gene product has been proposed to be an adapter or docking molecule. Mutations in this gene result in autosomal recessive Charcot-Marie-Tooth disease type 4C, a childhood-onset neurodegenerative disease characterized by demyelination of motor and sensory neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3TC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SH3TC2
Gene Accession No. Q80VA5, Q8TF17
Gene Alias CMT4C; D430044G18Rik; KIAA1985; MNMN; PP12494; RGD1309038; SH3 domain and tetratricopeptide repeat-containing protein 2; SH3 domain and tetratricopeptide repeats 2; SH3 domain and tetratricopeptide repeats-containing protein 2; SH3TC2
Gene Symbols SH3TC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SH3TC2(Accession Q8TF17), corresponding to amino acid residues L1244-L1288.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225608, 79628
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.