Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Double Curved Post-Mortem Needle, Double curved; 4.5 in. (11.4cm)

Catalog No. p-4520677 Shop All Epredia Products
Click to view available options
Length (Metric):
11.4 cm
12.7 cm

Epredia™ Shandon Double Curved Post-Mortem Needles

  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship

Specifications

Product Type Post-Mortem Needle, Double Curved
Tip Style Double Curved
Length (English) 5 in.
Length (Metric) 12.7 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.