Learn More
Epredia™ Embedding Cassettes in Tube Packs
Catalog No. p-4524770 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
This item is not returnable. View return policy
Catalog No. B1000732AQ
Supplier: Epredia™ B1000732AQ
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
  • Can be used on all Epredia PrintMate automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
  • Made of acetal polymer with a pore style of round holes
  • Supplied in 10 tubes of 75 cassettes each
Specifications
Pore Style Round Holes
Quantity 750 Cassettes/Cs.
Color Aqua
Material Acetal Polymer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.