Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Excelsior™ AS Tissue Processor Accessories

Catalog No. A78410026 Shop All Epredia Products
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Random cassette basket
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit

Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.

For use with Shandon Excelsior ES Tissue Processor

Specifications

Product Type Storage Bottle
For Use With (Equipment) Excelsior AS Tissue Processor
Quantity 5 L, 6/Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.