Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin

Catalog No. 9990244 Shop All Epredia Products
Click to view available options
Quantity:
120 mL container 60 mL filled
3.8 L
5 gal. (19L)
60 mL container 30 mL filled
90 mL
Reconstituted to 5 Liters (Ea.)
This item is not returnable. View return policy
This item is not returnable. View return policy

Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available

Specifications

Chemical Name or Material Phosphate-free neutral buffered 10% Formalin
Packaging 2/case
Quantity Reconstituted to 5 Liters (Ea.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.