Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Gill™ Hematoxylin

Catalog No. 6765006 Shop All Epredia Products
Click to view available options
Quantity:
4L
2 x 1L
Type:
Shandon Gill 1 Hematoxylin
Shandon Gill 2 Hematoxylin
Shandon Gill 3 Hematoxylin
This item is not returnable. View return policy
This item is not returnable. View return policy

Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.

  • Gill 1: Light blue nuclei

  • Gill 2: Medium blue nuclei

  • Gill 3: Blue-black nuclei

Specifications

Type Shandon Gill 1 Hematoxylin
Quantity 4L
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.