Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Kwik-Diff™ Stains

Catalog No. p-4530127
Click to view available options
Quantity:
125 mL
3 x 500 mL
4 L
Type:
Eosin
Fixative
Kwik-Diff Stains
Methylene Blue
Includes:
125 mL Each: Fixative, Eosin, Methylene Blue
4 L Kwik-Diff Reagent 1, Fixative
4 L Kwik-Diff Reagent 2, Eosin
4 L Kwik-Diff Reagent 3, Methylene Blue
500 mL Each: Fixative, Eosin, Methylene Blue
This item is not returnable. View return policy
This item is not returnable. View return policy

Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.

  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori

Includes:

Reagent 1: Fixative; Reagent 2: Eosin; Reagent 3: Methylene Blue

Specifications

Quantity 3 x 500 mL
Type Kwik-Diff Stains
Includes 500 mL Each: Fixative, Eosin, Methylene Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.