Learn More
Epredia™ Shandon™ Nu-Clear™ Reagent
Catalog No. 6769009 Shop All Epredia Products
Click to view available options
Product Type:
Tissue Processing Reagent (Nu-Clear I)
Tissue Processing Reagent (Nu-Clear II)
This item is not returnable. View return policy
This item is not returnable. View return policy
Achieve improved cell clarity by removing excess hematoxylin from the slide with Epredia™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.
Key Features 
  • Removes residual hematoxylin staining the glass.
  • Designed to be used on the full line of Epredia Shandon automatic stainers as well as manual methods.
Nu-Clear I
  • Fast-acting solution
  • Recommended to be used as a 10-second step in most protocols
  • Particularly beneficial for use with Harris hematoxylin
  • May be used on the Epredia Varistain™ 24-4 or Shandon Varistain Gemini slide stainers
Nu-Clear II
  • Less concentrated
  • Recommended as a 20-second step
  • May be used on the Epredia Shandon Linistat and other linear stainers
  • Rcommended for use with Gill hematoxylins
Specifications
Product Type Tissue Processing Reagent (Nu-Clear II)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.