Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
White
Yellow
Description
- Acetal polymer construction
- 30° writing face
- Small slot-style pores
- Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Specifications
| Pore Style | Slotted Pore |
| Writing Face Angle | 30° |
| Color | Yellow |
| Material | Acetal Polymer |
| Product Type | Tissue Cassette |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov