Learn More
Epredia™ Shandon™ Wright-Giemsa Stain Kit
Catalog No. p-6338600
Click to view available options
Quantity:
500 mL
Type:
Wright-Giemsa Stain
Eosinates of polychrome methylene blue yield rich color
  • Unique manufacturing process produces an intense differential stain for blood smears, bone marrows, mast cells, and FNA's
  • Demonstrates microorganisms such as Microfilaria, Babesia, Haemobartonella, Giardia, Pneumocystis, and Helicobacter
Specifications
Quantity 500 mL
Type Wright-Giemsa Stain
Includes Stain, buffer and rinse
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.