Learn More
Epredia™ Xylene
Catalog No. 9990501 Shop All Epredia Products
Click to view available options
Quantity:
1 Gallon, Ea.
1 Gallon; 4/Cs.
5 liter; 4/Cs.
55 gallon; steel drum
This item is not returnable. View return policy
This item is not returnable. View return policy
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with this Xylene.
  • For use in all tissue processors and automatic stainers
  • Contains the lowest level of benzene, a known carcinogen
  • Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product
Specifications
Product Type Xylene
Quantity 1 Gallon, Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.