Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHANK1 Monoclonal Antibody (N22/21), PerCP
GREENER_CHOICE

Catalog No. PIMA545448
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545448 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545448 Supplier Invitrogen™ Supplier No. MA545448
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45448 was sufficient for detection of Shank1 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 190-220 kDa (alternative splice variants). No cross-reactivity against Shank2 or Shank3.

Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. May play a role in the structural and functional organization of the dendritic spine and synaptic junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHANK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N22/21
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SHANK1
Gene Accession No. D3YZU1, Q9WV48, Q9Y566
Gene Alias GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; OTTHUMP00000174437; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3/ankyrin domain gene 1; SHANK 1; Shank1; Shank1a; somatostatin receptor-interacting protein; SPANK 1; Spank1; SPANK-1; SSTR-interacting protein; Sstrip; Synamon
Gene Symbols SHANK1
Host Species Mouse
Immunogen Fusion protein amino acids 469-691 (SH3/PDZ domains) of rat Shank1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243961, 50944, 78957
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.