Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHANK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14180 Supplier Invitrogen™ Supplier No. PA14180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4180 detects SHANK1a in human, rat, and mouse samples. PA1-4180 has been successfully used in Western blot, immunofluorescence, and immunofluoresence procedures. By Western blot this antibody detects a ~212 kDa protein representing SHANK1a in rat brain lysate. The PA1-4180 immunogen is a synthetic peptide corresponding to the C-terminal residues S(1951) G P I Y P G L F D I R S S(1964) of rat Shank1a. Maintain at 4ºC for 3 months or at -20ºC for longer periods. Avoid freeze/thaw cycles.

Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. May play a role in the structural and functional organization of the dendritic spine and synaptic junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHANK1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SHANK1
Gene Accession No. D3YZU1, Q9WV48, Q9Y566
Gene Alias GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; OTTHUMP00000174437; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3/ankyrin domain gene 1; SHANK 1; Shank1; Shank1a; somatostatin receptor-interacting protein; SPANK 1; Spank1; SPANK-1; SSTR-interacting protein; Sstrip; Synamon
Gene Symbols SHANK1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues S(1951) G P I Y P G L F D I R S S(1964) of rat Shank1a.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243961, 50944, 78957
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.