Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shank2 Monoclonal Antibody (S23b-6), QED Bioscience™
SDP

Catalog No. 502278195
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8195 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8195 Supplier Leinco Technologies Inc Supplier No. 56466100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Shank2 Monoclonal antibody specifically detects Shank2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen Shank2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S23b-6
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene SHANK2
Gene Accession No. Q80Z38, Q9QX74, Q9UPX8
Gene Alias AUTS17; cortactin SH3 domain-binding protein; cortactin-binding protein 1; CortBP1; CORTBP1CTTNBP1; CTTNBP1; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; Kiaa1022; mKIAA1022; proline rich synapse associated protein 1; proline-rich synapse associated protein 1; proline-rich synapse-associated protein 1; PROSAP1; SH3 and multiple ankyrin repeat domains 2; SH3 and multiple ankyrin repeat domains protein 2; SH3/ankyrin domain gene 2; SHANK; Shank2; SPANK-3
Host Species Mouse
Immunogen Fusion protein corresponding to aa 84-309 (SH3/PDZ domains) of rat Shank2 (accession no. Q9QX74). This sequence is 94
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171093, 210274, 22941
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.