Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHANK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578652 Supplier Invitrogen™ Supplier No. PA578652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted Reactivity: Human (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is a member of the Shank family of synaptic proteins that may function as molecular scaffolds in the postsynaptic density of excitatory synapses. Shank proteins contain multiple domains for protein-protein interaction, including ankyrin repeats, and an SH3 domain. This particular family member contains a PDZ domain, a consensus sequence for cortactin SH3 domain-binding peptides and a sterile alpha motif. The alternative splicing demonstrated in Shank genes has been suggested as a mechanism for regulating the molecular structure of Shank and the spectrum of Shank-interacting proteins in the postsynaptic densities of the adult and developing brain. Alterations in the encoded protein may be associated with susceptibility to autism spectrum disorder. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHANK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene SHANK2
Gene Accession No. Q80Z38, Q9QX74
Gene Alias AUTS17; cortactin SH3 domain-binding protein; cortactin-binding protein 1; CortBP1; CORTBP1CTTNBP1; CTTNBP1; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; Kiaa1022; mKIAA1022; proline rich synapse associated protein 1; proline-rich synapse associated protein 1; proline-rich synapse-associated protein 1; PROSAP1; SH3 and multiple ankyrin repeat domains 2; SH3 and multiple ankyrin repeat domains protein 2; SH3/ankyrin domain gene 2; SHANK; Shank2; SPANK-3
Gene Symbols SHANK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse SHANK2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171093, 210274
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.