Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHANK2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | SHANK2 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SHANK2 Polyclonal antibody specifically detects SHANK2 in Human samples. It is validated for ImmunofluorescenceSpecifications
SHANK2 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.2, 40% glycerol | |
22941 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
AUTS17, cortactin binding protein 1, cortactin SH3 domain-binding protein, Cortactin-binding protein 1, CortBP1, CORTBP1CTTNBP1, GKAP/SAPAP interacting protein, KIAA1022, proline-rich synapse associated protein 1, Proline-rich synapse-associated protein 1, PROSAP1, SH3 and multiple ankyrin repeat domains 2, SH3 and multiple ankyrin repeat domains protein 2, SHANK, Shank2, SPANK-3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DRAMKESQQGPKGEAPKADLNKPLYIDTKMRPSLDAGFPTVTRQNTRGPLRRQETENKYETDLGRDRKGDDKKNMLIDIMDTSQQKSAGLLMVHTVDATKLDNALQEEDEKAEVEMKPDSSPSEVPEGVSETEGALQ | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title