Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHANK3 Antibody (S69), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147610H
Description
SHANK3 Monoclonal specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SHANK3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 190kDa. No cross-reactivity against Shank1 or Shank2. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S69-46 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q9JLU4 | |
SHANK3 | |
Synthetic peptide amino acids 840-857 of rat Shank3 | |
0.1 mL | |
85358 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction